Pilot Remote software
Lieferant: HUBER
10645
HUBE10645EA 0 EUR
HUBE10645
Pilot Remote software
Software Labor-Software
The Pilot Remote software enables the complete remote control of temperature control units with Pilot ONE through a Windows PC.
  • Remote control of any number of temperature control units with Pilot ONE
  • Secure Ethernet connection with authentication
  • Compatible with Microsoft Windows 7 / 8 / 10 (32 / 64 bit)
The user display of the Pilot ONE is equally displayed on the PC, this means identical operation on PC and temperature control unit. The communication takes place via Ethernet network connectivity with a secure authentication and encoding.
Lieferumfang: Supplied with hardware dongle (for any Windows PC).