Thermostatic baths for viscometers, CC®
Lieferant: HUBER
2001.0006.01 2048.0001.01
HUBE2001.0006.01EA 3640 EUR
HUBE2001.0006.01 HUBE2048.0001.01
Thermostatic baths for viscometers, CC®
Bäder Viskometer-Bäder
Visco thermostats are mainly used with capillary viscometers or densitometers for the measurement applications. These are offers the highest temperature stability for optimal temperature distribution in the thermostat bath.
  • Pilot ONE controller with plug and play technology
  • Precise temperature control
  • Overtemperature and low-level protection
  • Pt100 sensor connection
  • Powerful pressure/suction pump
  • Two-point calibration
The units offer the highest temperature stability for optimal temperature distribution in the thermostat bath. For operation below or near the ambient temperature there is a cooling coil integrated for a cooling water connection. Various function packages for the Pilot ONE can be activated using E-grade as required. The new multi-touch Pilot ONE controller offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu. All-important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favorites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
Maximum pump capacities: 27 L/min, 700 mbar.
Standards: Safety class III/FL (DIN 12876).