Wasserbäder, Edelstahl, für Kühlung
Lieferant: HUBER
2009.0032.00 2010.0026.00 2011.0022.00 2012.0026.00
HUBE2009.0032.00EA 3540 EUR
HUBE2009.0032.00 HUBE2010.0026.00 HUBE2011.0022.00 HUBE2012.0026.00
Wasserbäder, Edelstahl, für Kühlung
Bäder Wasserbäder, ohne Umwälzung
The Cooling Bath are water baths with air-cooled refrigerating unit and use natural refrigerants. Housing and tank of stainless steel. The refrigerating unit works continuously.
In combination with an immersion circulator these refrigerated bath systems offers active cooling, in continuous operation over the complete working range.
Bitte beachten Cooling capacity at +20; 0; −10; −20; −30 °C.