Heating circulation thermostats, Hotbox
Lieferant: HUBER
2030.0001.01 2031.0004.01
HUBE2030.0001.01EA 11350 EUR
HUBE2030.0001.01 HUBE2031.0004.01
Heating circulation thermostats, Hotbox
Umwälzthermostate Geschlossene Umwälzsysteme Wärmeumwälzthermostate
These compact hotbox heating circulators are ideal for installation in production systems. All models have self-optimising True Adaptive Control for temperature. Units feature a PT100 sensor connection, digital filling level indicator, and circulation pump. All models feature the Pilot ONE® controller which offers a brilliant 14,5 cm touchscreen with graphic functions and a comfortable navigation menu in 11 languages and USB and Ethernet connections.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • Powerful circulation pump
  • All important operating parameters are clearly displayed on the touchscreen
  • All models can be used for open circulating applications
Maximum pump capacities:
100 L/min, 250 mbar pressure
Standards: Comply with DIN 12876 Safety class II, FL