Software for huber units
Lieferant: HUBER
6792
HUBE6792EA 0 EUR
HUBE6792
Software for huber units
Software Labor-Software
SpyControl® software enables process relevant data to be visualised and documented.
  • Communication options: RS232, RS485, USB (virtual COM-Port) or TCP/IP
  • Easy to install
  • Recorded data is displayed to a base of time
The software can operate up to 10 channels simultaneously. Each channel is independently documented and the graphic options can be configured as required. SpyControl allows the user to issue the following instructions to the unit such as set point, change from jacket to process temperature and start or stop. The software offers the option of controlling one or more machines with a programmer. The user can provide temperature programs for the machines which then automatically run.