Breather controller for unistats
Lieferant: HUBER
9771
HUBE9771EA 0 EUR
HUBE9771
Breather controller for unistats
Regler Utility-Controller
This breather controller enables small pressurisation of the expansion vessel.
  • Breather control limits the pressure for the pressure overlay in the expansion vessel
  • Expansion vessel in the unistats can handle a pressure of up to 0,3 bar
  • Prolonges life-time of the thermal fluid due to the reduced oxidation as well as an extended temperature range
The breather control consists of the atmospheric sealing kit and the parts built onto the mounting plate: Pressure regulator (20 – 200 mbar), needle valve, pressure relief valve, bubble counter and connection hose. The pressure regulator does have a preset pressure relief valve that will blow-off at too high a pressure.
Bitte beachten Not suitable for outdoor installation – Do not install in a closed room. Room must be adequately ventilated.