Spigot with screw caps for jerricans
Lieferant: hünersdorff GmbH
550500
HUEN550500EA 73.9 EUR
HUEN550500
Spigot with screw caps for jerricans
Hähne
Spigots PROFI with red DIN 60/61 screw cap is ideal for controlled emptying of liquids from industrial jerrycans.
  • Resistant to acids, alkalis and other chemicals
  • Pressure resistance up to 0,4 bar