Heating oil cans, with sieve
Lieferant: hünersdorff GmbH
821200
HUEN821200EA 46.4 EUR
HUEN821200
Heating oil cans, with sieve
Flaschen Ballons
This oil can, made of HDPE, grey, could be used in industry, workshop and household.
  • Have a sieve insert at the filling opening
  • With screw cap at the outlet
  • Resistant to oil, diesel, petrol and diluted acids