Dulbecco's phosphate buffered saline (DPBS), for cell culture, powder format, HyClone™
Lieferant: HyClone products (Cytiva)
SH30013.04
SH30013.02
SH30013.03
HYCLSH30013.04EA
98.3
EUR
HYCLSH30013.04
HYCLSH30013.02
HYCLSH30013.03
Dulbecco's phosphate buffered saline (DPBS), for cell culture, powder format, HyClone™
Puffer
Gewebekultur-Puffer
Balanced salt solution used for multiple cell culture applications (powder form).
- Formulated and processed for rapid dissolution
- Non sterile
- Pre-weighed powders ready for hydration to 1X
- Available in 3" ported funnel shaped containers upon request
- Available in lot sizes up to 6500 kg.
For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.
Standards: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Bitte beachten Not for diagnostic or therapeutic use.