HyClone™ FetalClone™ I serum, U.S. origin
Lieferant: HyClone products (Cytiva)
SH30080.02 SH30080.03 SH30080.02HI SH30080.03HI SH30080.03IR25
SH30080.02EA 153 EUR
SH30080.02 HYCLSH30080.03 HYCLSH30080.02HI HYCLSH30080.03HI SH30080.03IR25
HyClone™ FetalClone™ I serum, U.S. origin
Seren
HyClone™ FetalClone® I is a foetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
  • IgG levels comparable with those found in FBS
  • Demonstrated cell growth performance equivalency to FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.