Wasser, HyClone™ HyPure zur Injizierung (WFI), steril gefiltert

Lieferant: HyClone products (Cytiva)

SH30221.25 SH30221.17 SH30221.28
HYCLSH30221.25EA 931 EUR
HYCLSH30221.25 HYCLSH30221.17 HYCLSH30221.28
Wasser, HyClone™ HyPure zur Injizierung (WFI), steril gefiltert
Wasser

Water for Injection (WFI) quality water supports cell culture growth and biomanufacturing processes.


  • Meets stringent USP specifications
  • Manufactured in ISO 9001 certified facility
  • Lot-specific certificate of analysis (CoA) for each lot of water produced
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use
  • 0,1 µm sterile filtered


HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of the United States Pharmacopeia (USP).


HyPure WFI Quality Water meets the following criteria: Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water. Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits. Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water. Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results. Actual test results on a CoA are not taken from system samples, but must be from actual packaged samples of the given lot.


Standards: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.


Bestellinformation: All purified waters are supplied in PETE/PETG bottles, except for HYCLSH30221.24, HYCLSH30221.26 and HYCLSH30221.27, each of which is supplied in a Thermo Scientific HyClone® BioProcess Container (BPC®) system. For alternate volumes and container options, contact your VWR sales representative.

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Sterilisation 0,1 µm sterile filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR