Trypsin Protease, HyClone™
Lieferant: HyClone products (Cytiva)
SH30236.01 SH30236.02 SH30042.01 SH30042.02 SV30031.01 SV30037.01
HYCLSH30236.01EA 24.9 EUR
HYCLSH30236.01 HYCLSH30236.02 SH30042.01 HYCLSH30042.02 HYCLSV30031.01 HYCLSV30037.01
Trypsin Protease, HyClone™
Zellkultur-Ergänzungen und -Zusätze
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium