HEPES Buffer Solution 1 M, HyClone™

Lieferant: HyClone products (Cytiva)

SH30237.01
HYCLSH30237.01EA 43.2 EUR
HYCLSH30237.01
HEPES Buffer Solution 1 M, HyClone™
Puffer Gewebekultur-Puffer

Maintain physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration) with Cytiva HyClone HEPES solution.


  • 0,1 µm sterile filtered
  • pH: 7,2 - 7,5
  • Storage: 2 to 8° C


Standards: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc04763bdb604a22b88a12360e93b1d1b17277e0&vid=a6e236e4cd25af39631a515bbad5e3397cbd64bf&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR