Newborn bovine calf serum, New Zealand origin, HyClone™
Lieferant: HyClone products (Cytiva)
SH30401.01 SH30401.02 SH30401.01HI SH30401.02HI
HYCLSH30401.01EA 132 EUR
HYCLSH30401.01 HYCLSH30401.02 HYCLSH30401.01HI HYCLSH30401.02HI
Newborn bovine calf serum, New Zealand origin, HyClone™
Seren
Newborn bovine calf serum is a cost-effective alternative to foetal bovine serum (FBS). The serum is sourced from animals that are typically less than 10 days old.