Wasser, HyClone™ HyPure, Zellkultur-Qualität

Lieferant: HyClone products (Cytiva)

SH30529.01 SH30529.02 SH30529.03 SH30529.FS SH30529.LS
HYCLSH30529.01EA 14.4 EUR
HYCLSH30529.01 HYCLSH30529.02 SH30529.03 HYCLSH30529.FS HYCLSH30529.LS
Wasser, HyClone™ HyPure, Zellkultur-Qualität
Wasser

Eliminate endotoxin concerns with endotoxin-free HyClone Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.


  • Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP)
  • Meets < 0,005 EU/mL endotoxin by turbidimetric assay
  • Manufactured in our ISO 9001 certified facilities


HyClone HyPure Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0,005 EU/mL, HyPure Cell Culture Grade Water supports healthy cell growth and consistent experimental results.


Standards: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Endotoxin Concentration < 0,005 EU/mL
Sterilisation 0,1 µm sterile filtered
pH 5,0 to 7,0
Storage 2° to 30° C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR