L-alanyl L-glutamine, SG-200, HyClone™
Lieferant: HyClone products (Cytiva)
SH30590.01
HYCLSH30590.01EA
17.1
EUR
HYCLSH30590.01
L-alanyl L-glutamine, SG-200, HyClone™
Zellkultur-Ergänzungen und -Zusätze
SG-200 solution is a stable dipeptide of L-alanyl L-glutamine.