Penicillin/Streptomycin/Fungizone-Lösung 100X, HyClone™

Lieferant: HyClone products (Cytiva)

SV30079.01
HYCLSV30079.01EA 38.2 EUR
HYCLSV30079.01
Penicillin/Streptomycin/Fungizone-Lösung 100X, HyClone™
Penicillin/Streptomycin/Fungizone

Antibiotic-Antimycotic (Pen/Strep/Fungiezone) Solution is used to control or destroy destructive microorganisms, such as gram negative, gram positive bacteria, fungi and yeast.


  • Contains 10 000 units of penicillin, 10 000μg of streptomycin, and 25 μg of Amphotericin B per ml
  • Wide spectrum of activity against gram negative and gram positive bacteria, as well as activity against fungi and yeast
  • Commonly used as an anti-fungal, anti-yeast, anti-mould or anti-bacterial
  • Common working dose is 10 ml/l of 100X solution

Bestellen

Spezifikation Testergebnisse

Format Liquid
Storage Store frozen
Antibiotic Spectrum Broad spectrum antibacterial and antifungal
Concentration 100X
Shelf life 18 months
Sterilization 0.2 µm sterile filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR