SMART Digest™ Chymotrypsin Kits
Lieferant: Thermo Fisher Scientific
60109-104 60109-105 60109-106 60109-104-B 60109-104-MB 60109-105-B 60109-105-MB 60109-106-B 60109-106-MB
555-0125EA 963 EUR
555-0125 555-0126 555-0127 HYPE60109-104-B HYPE60109-104-MB HYPE60109-105-B HYPE60109-105-MB HYPE60109-106-B HYPE60109-106-MB
SMART Digest™ Chymotrypsin Kits
SMART Digest™ Chymotrypsin Kits are designed to provide a complimentary digestion option to the SMART Digest™ Trypsin Kits which incorporate trypsin functionality. The Chymotrypsin enzyme functionality allows digestion of proteins at different points of the amino acid chain compared to trypsin. This allows for different/additional structural information to be obtained when trying to characterise a protein. This option can also be used if a protein does not contain the specific point where trypsin cuts the protein chain.
  • Significantly faster than in-solution digestion
  • Simple process
  • High reproducibility
  • Amenable to high-throughput processing
Lieferumfang: Available as a stand alone kit, or with optional filter plate or SOLAµ™ SPE plate. Bulk resin (-B) and magnetic bulk resin (-MB) are also available separately.