Electronic hand-held crimpers and decrimpers, level 2 high throughput applications, SureSTART™
Lieferant: Thermo Fisher Scientific
60180-ECR11 60180-ECR13 60180-ECR20 60180-ECR8 60180-ECRHB 60180-EDCB11 60180-EDCB13 60180-EDCB20
HYPE60180-ECR11EA 1480 EUR
HYPE60180-ECR11 HYPE60180-ECR13 HYPE60180-ECR20 HYPE60180-ECR8 HYPE60180-ECRHB HYPE60180-EDCB11 HYPE60180-EDCB13 HYPE60180-EDCB20
Electronic hand-held crimpers and decrimpers, level 2 high throughput applications, SureSTART™
Crimper, Capper und Decapper
Use Thermo Scientific™ electronic hand-held vial crimpers and decrimpers to attach or remove crimp caps from vials, respectively.
These hand-held, high-power crimpers and decrimpers are available in 8, 11, 13, and 20 mm sizes and allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps. The electronic crimper's message window conveniently provides all-important information at one glance. This device is performance level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.