GFM Pro Electronic Gas Flow Meter
Lieferant: Thermo Fisher Scientific
66002-010
HYPE66002-010EA 1380 EUR
HYPE66002-010
GFM Pro Electronic Gas Flow Meter
Durchflussmesser
The Thermo Scientific GFM Pro Flowmeter is specifically designed for use with gas chromatography instruments.
  • Data output via USB port
  • Uses two AA batteries
  • Recalibration service available
  • Easy to use interface features an over-range warning indicator
  • Automatic shutoff and explosion-proof rating for use with flammable and explosive gases
  • Compact ergonomic design enables use as a handheld or benchtop unit and features side grips for added durability
This versatile product is an electronic device capable of measuring volumetric flow for all types of gases. Real-time measurements can be made for various types of flow paths including continually changing gas types.
Standards: Calibration traceable to NIST primary standards.