SureSTART™ Specification Certified Vial and Cap Kits, Level 1
Lieferant: Thermo Fisher Scientific
6EKSHV8-2
HYPE6EKSHV8-2EA 59.1 EUR
HYPE6EKSHV8-2
SureSTART™ Specification Certified Vial and Cap Kits, Level 1
Vials
Select Level 1 products for an affordable option for everyday chromatography analysis using core detector technology.
  • For everyday analysis
  • Chromatography approved
  • Not autoclavable
  • No preservative
  • Shell vials
Choose from a variety of kits containing short thread screw vials in clear propylene and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues.
SureSTART Level 1 products are cost optimised, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.