Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Immersion oil has been used in epifluorescent microscopy and cell identification.
- Suitable for Hematology
- Suitable for Histology
Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.