L(-)-Tyrosin
Lieferant: IBS Stock Items
L-TYROSIN
IBSAL-TYROSINEA
0
EUR
IBSAL-TYROSIN
L(-)-Tyrosin
L(-)-Tyrosin
Formel:
C₉H₁₁NO₃ Siedepunkt: 351 °C (1013 hPa) Schmelzpunkt: 285 °C Dichte: 1,41 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002606 CAS-Nummer: 60-18-4 EINECS: 200-460-4 Merck Index: 13,09907 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...