Aluminiumoxid, Ecochrom™ Alumina B Activity: I

Lieferant: MP Biomedicals

0202069
ICNA0202069EA 40.4 EUR
ICNA0202069
Aluminiumoxid, Ecochrom™ Alumina B Activity: I
Aluminium oxide

Specific Surface Area: 150 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 10


pH: 10

Formel: Al₂O₃
Dichte: 3,94 g/cm³ (20 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR