Aluminiumoxid, Ecochrom™ Alumina N Activity: I

Lieferant: MP Biomedicals

Synonyme: Aluminium(III) oxide, Alumina C, Aluminum oxide

0202084
ICNA0202084EA 40.4 EUR
ICNA0202084
Aluminiumoxid, Ecochrom™ Alumina N Activity: I
Aluminium oxide

Specific Surface Area: 150 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 7.5


pH: 7.5

Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR