Aluminiumoxid, Ecochrom™ Alumina N Activity: I
Lieferant: MP Biomedicals
Synonyme:
Aluminium(III) oxide, Alumina C, Aluminum oxide
0202090
ICNA0202090EA
298
EUR
ICNA0202090
Aluminiumoxid, Ecochrom™ Alumina N Activity: I
Aluminium oxide
Specific Surface Area: 150 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 7.5
pH: 7.5
Formel:
Al₂O₃ Molecular Weight: 101.96 g/mol Siedepunkt: 2980 Schmelzpunkt: 2045 Dichte: 3.94 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003424 CAS-Nummer: 1344-28-1 Merck Index: 13,00355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...