Siliciumdioxid, Ecochrom™ 32-63 µm
Lieferant: MP Biomedicals
Synonyme:
Silicium(IV)oxid
0202827
ICNA0202827EA
181
EUR
ICNA0202827
Siliciumdioxid, Ecochrom™ 32-63 µm
Siliciumdioxid
- Specific Surface Area: 500-600 m2/g
- Particle Size: 32-63 µm
- Weight: 0.4-0.5 g/ml
- pH: 7.0
- Mean Pore Size: 60Å
pH: 7
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 Schmelzpunkt: 1719 Dichte: ∼2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 7631-86-9 Merck Index: 14,08493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...