Aluminiumoxid basisch, MP Ecochrom™ Super I

Lieferant: MP Biomedicals

0204568
ICNA0204568EA 43.8 EUR
ICNA0204568
Aluminiumoxid basisch, MP Ecochrom™ Super I
Aluminiumoxid basisch

MP EcoChrom™ Alumina B, Activity Super I available as basic alumina is commonly used for purification processes of all kinds, including sample preparation for pesticides, and environmental analysis, solvent purification and all kinds of chromatography, requiring the highest precision and reproducibility.


  • Specific Surface Area: 200 m2/g
  • Particle Size: 63-200 µm
  • Weight: 0.8 g/ml
  • pH: 10


pH: 10

Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
Siedepunkt: ∼2980 °C
Schmelzpunkt: ∼, 2072 °C
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR