Alumina B, Activity Super I, weiß to hell rosa frei rinnendes Pulver, MP EcoChrom™ zur Dioxinanalyse

Lieferant: MP Biomedicals

0204569
ICNA0204569EA 51.8 EUR
ICNA0204569
Alumina B, Activity Super I, weiß to hell rosa frei rinnendes Pulver, MP EcoChrom™ zur Dioxinanalyse
Alumina B, activity super I

MP EcoChrom™ Alumina, Activity Super I is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This product meets the high recovery level required for the analysis of polychlorinated dibenzodioxins and dibenzofurans according to VDI Regulation 3499, part I.
MP EcoChrom™ Alumina B, Activity Super I is used for Dioxin analysis. It also been used for all chromatographic processes requiring highest activity and/or highest reproducibility.
Store at Room Temperature (15-30 °C)


  • Grade: for Dioxin Analysis
  • Presentation: White to Light Pink Free Flowing Powder
  • Weight: 0.8g/mL
  • Surface Area: 200 m2/g
  • Particle Size: 63-200 μm
  • Insoluble in water


pH: 10

Formel: Al₂O₃
Siedepunkt: ∼2980 °C
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
pH 9.3-10.1
Activity acc to Brockmann 14 - 16 l/mm
Loss of Ignition 0.6-0.8%
Particle Size Distribution Fine Parts <63 µm ≤10%
Particle Size Distribution Large Parts >200 µm ≤5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR