Siliciumdioxid
Lieferant: MP Biomedicals
Synonyme:
Silicium(IV)oxid
0204660
ICNA0204660EA
1862
EUR
ICNA0204660
Siliciumdioxid
Siliciumdioxid
Specific Surface Area: 500-600 m2/g
Particle Size: 63-100 µm
Weight: 0.4-0.5 g/ml pH: 7.0
Formel:
SiO₂ Molecular Weight: 60,08 g/mol Dichte: 2,65 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 7631-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...