Aluminiumoxid, sauer, MP Ecochrom™, technische Qualität A

Lieferant: MP Biomedicals

0207895
ICNA0207895EA 0 EUR
ICNA0207895
Aluminiumoxid, sauer, MP Ecochrom™, technische Qualität A
Aluminiumoxid saures

MP EcoChrom TM aluminum oxide, acid technical A is used for isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones and antibiotics, dehydration of organic solvents, purification of enzymes, removal of terpenes, peroxides, pyrogens from injectable solutions and infusions.

Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR