N-Alpha-Benzoyl-L-Argininamid-Hydrochlorid-Monohydrat, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: Nardis Reagent, Bz-Arg-NH2 HCl, BAA, N-α-Benzoyl-L-argininamide hydrochloride monohydrate

0210008705
ICNA0210008705EA 300 EUR
ICNA0210008705
N-Alpha-Benzoyl-L-Argininamid-Hydrochlorid-Monohydrat, weißes Pulver
N-Alpha-Benzoyl-L-Argininamid-Hydrochlorid-Monohydrat

Enzyme substrate for papain.


Substrate for papain, chymopapain and papaya proteases.


Soluble in methanol (50 mg/ml-clear, colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₃H₁₉N₅O₂·HCl·H₂O
Molecular Weight: 313,79 g/mol (anhy)
MDL: MFCD00035003
CAS-Nummer: 4299-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR