N-Acetyl-DL-Tryptophan ∼99%, weißes Pulver
Lieferant: MP Biomedicals
Synonyme:
Ac-DL-Trp-OH, N(α)-Acetyl-DL-Tryptophan
0210014225
0210014280
0210014291
ICNA0210014225EA
59.9
EUR
ICNA0210014225
ICNA0210014280
ICNA0210014291
N-Acetyl-DL-Tryptophan ∼99%, weißes Pulver
N-Acetyl-DL-Tryptophan
N-Acetyl-DL-tryptophan, is used as stabiliser in the human blood-derived therapeutic products normal serum albumin and plasma protein fraction.
Clear colorless to light yellow solution at 25 mg/ml in 1N sodium hydroxide.
Formel:
C₁₃H₁₄N₂O₃ Molecular Weight: 246.27 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005644 CAS-Nummer: 87-32-1 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ~99% |
FTIR | Conforms to Standard |
Melting Point | 200 - 210°C |
Loss on Drying | ≤0.5% |
UV/VIS: λmax (water) | 280 ± 2 n |
Optical Rotation (c=1, 1N NaOH, 24 hrs.) | 0° ± 2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...