5-Iod-2'-deoxyuridin (Idoxuridin) ≥99%, kristallin
Lieferant: MP Biomedicals
|
Achtung
|
02100357.2
0210035701
0210035705
0210035725
ICNA02100357.2EA
46.1
EUR
ICNA02100357.2
ICNA0210035701
ICNA0210035705
ICNA0210035725
5-Iod-2'-deoxyuridin (Idoxuridin) ≥99%, kristallin
5-Iod-2'-deoxyuridin (Idoxuridin)
Storage temperature: -20 °C
Formel:
C₉H₁₁IN₂O₅ |
MDL:
MFCD00134656 CAS-Nummer: 54-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...