β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) ≥98%

Lieferant: MP Biomedicals

02100499.1 0210049901 0210049905 0210049910
ICNA02100499.1EA 53.2 EUR
ICNA02100499.1 ICNA0210049901 ICNA0210049905 ICNA0210049910
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) ≥98%
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)

Major electron acceptor molecule in biological oxidations. Acts as a coenzyme for hydride-transfer enzymes and a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins. Recently reported to be a P2Y11 agonist.


  • Lyophilised white powder
  • High purity


Storage temperature: –20 °C


pH: 2 to 4 (1% aquous solution)

Formel: C₂₁H₂₇N₇O₁₄P₂
MDL: MFCD00036253
CAS-Nummer: 53-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR