Puromycin Dihydrochlorid

Lieferant: MP Biomedicals

Synonyme: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

0210055210 0210055225 0210055280 0210055283 0210055290 0210055291
ICNA0210055210EA 44.7 EUR
ICNA0210055210 ICNA0210055225 ICNA0210055280 ICNA0210055283 ICNA0210055290 ICNA0210055291
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid

Puromycin is nucleoside antibiotic & protein synthesis inhibitor that causes premature chain termination by acting as an analogue of the 3′-terminal end of aminoacyl-tRNA. Prevents growth of bacteria, protozoa, algae, and mammalian cells.


Puromycin is an antibiotic of unique structure and biological activity. It is produced by fermentation using Streptomyces albo-niger. There are four aspects of puromycin's biological activity that are 1) anti tumour effect; 2) nephrotoxic action; 3) inhibition of purine and/or protein synthesis and 4) antitrypanosome activity.


Soluble in water (50 mg/ml - clear, colorless solution).

Formel: C₂₂H₂₉N₇O₅·2HCl
MDL: MFCD00012691
CAS-Nummer: 58-58-2

Bestellen

Spezifikation Testergebnisse

Synonyms 6-dimethylamino-9-(3'-deoxy-3'-(p-methoxy-L-phenylalanyamino)- B-D-ribofuranosyl)-purine
Form Off-white Powder
Storage Store at -0 °C
Extinction Coefficient 20,3 (275 nm; 0,1 N NaOH); 19,5 (268 nm, 0,1 NHCl) (Lit.)
pka 6,8; 7,2 (Lit.)
Source Streptomyces albo-niger


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR