Polymyxin B-Sulfat
Lieferant: MP Biomedicals
0210056550
0210056501
0210056505
0210056510
0210056525
ICNA0210056550EA
816
EUR
ICNA0210056550
ICNA0210056501
ICNA0210056505
ICNA0210056510
ICNA0210056525
Polymyxin B-Sulfat
Polymyxin B sulphate
Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.
Formel:
C₅₅H₉₆N₁₆O₁₃·2H₂SO₄ |
MDL:
MFCD00131991 CAS-Nummer: 1405-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...