Glycin (H-Gly-OH) 98, powder

Lieferant: MP Biomedicals

Synonyme: H-Gly-OH, Glycocoll, Aminoessigsäure

0210057025 02100570.5 0210057001 0210057005
ICNA0210057025EA 1310 EUR
ICNA0210057025 ICNA02100570.5 ICNA0210057001 ICNA0210057005
Glycin (H-Gly-OH) 98, powder
Glycin (H-Gly-OH)

Glycine is a non-essential amino acid. Acts as an inhibitory neurotransmitter, particularly in brain stem and medulla. Shown to block NMDA-stimulated dopamine release in fetal mesencephalic cell cultures. Also reported to increase NMDA-receptor mediated synaptic potential in cortical neurons. It does not have a chiral center, so only amino acid with no asymmetric carbon. Therefore, it does not have a D- or L- isomer designation. This material is synthetic. It is the smallest of the 20 natural amino acids.


Glycine is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments. The growth requirements of various microorganisms is reported in the Handbook of Microbiology.


It is a non-chiral amino acid that can be synthesised in the body from the amino acid serine by Serine Hydroxy methyltransferase.


Soluble in water (250 mg/ml at 25 °C; 672 mg/ml at 100 °C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.


Soluble in water (250 mg/ml at 25 °C; 672 mg/ml at 100 °C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.

Formel: NH₂CH₂COOH
Molecular Weight: 75.07 g/mol
Siedepunkt: 181
Schmelzpunkt: 83
Dichte: 1.64 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Synonyms Amino acetic acid; Gly; Amino ethanoic acid; Glycocoll
Form Powder
Storage Store at room temperature (15 to 30 °C)
Isoelectric Point 6,06
Loss on Drying ≤0,5%
pka for the α carboxyl group: 2,35; and for the α amino group: 9,78 at 25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR