1,4-Dithioerythrit ≥99%, weißes kristallines Pulver

Lieferant: MP Biomedicals
Achtung

02100598.5 0210059825 0210059801 0210059805 0210059810 0210059880
ICNA02100598.5EA 51.4 EUR
ICNA02100598.5 ICNA0210059825 ICNA0210059801 ICNA0210059805 ICNA0210059810 ICNA0210059880
1,4-Dithioerythrit ≥99%, weißes kristallines Pulver
1,4-Dithioerythrit

Dithioerythritol is an erythro isomer of 2,3-dihydroxy-1,4-dithiolbutane, and an isomer of 1,4-dithiothreitol which is a reagent for maintaining thiols in the reduced state; prevents oxidation of sulfhydryl-containing proteins during SDS-polyacrylamide gel electrophoresis. It also quantitatively reduces disulfides.


Dithioerythritol is used to prepare a matrix for fast-atom bombardment mass spectroscopy. It has been reported to be used in 2-D electrophoretic analysis of proteins and immunoglobulins, mitochondrial studies to block the opening of the transition pore and to study permeability transition, glial and neuronal glycine transporters have been investigated, utilised in a mass spectrometric mapping study of disulphide bonds in recombinant human interleukin-13.


Soluble in water (50 mg/ml - clear to slightly hazy, colourless solution).


Bitte beachten Store at +4 °C, protect from light, store under nitrogen

Formel: C₄H₁₀O₂S₂
Siedepunkt: 286 °C (1013 hPa)
Schmelzpunkt: 82 - 83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063750
CAS-Nummer: 6892-68-8
EINECS: 229-998-8
UN: 3335
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference
Melting Point 80 - 90 °C
UV/ VIS: OD283 (0,02 M aq soln) ≤0,04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR