L(+)-Arginin freie Base ≥99%, kristallin

Lieferant: MP Biomedicals

0210073680 0210073690 0210073691
ICNA0210073680EA 60.5 EUR
ICNA0210073680 ICNA0210073690 ICNA0210073691
L(+)-Arginin freie Base ≥99%, kristallin
L(+)-Arginin

L-Arginine is one of the three amino acids with basic side chains, and is very hydrophilic in character. It contains a guanidino group in the side chain, and this moiety is protonated at physiological pH.


Arginine is biosynthesized in the kidneys from citrulline, whose precursor is glutamate via the formation of ornithine. In amino acid degradation in vivo, arginine is hydrolyzed to urea and ornithine by arginase. Arginine can be metabolized to glutamate, which in turn is converted to α-ketoglutarate for entry into the citric acid cycle.


L-Arginine plays an important role in cell division, the healing of wounds, removing ammonia from the body, immune function, and the release of hormones. It is used as a precursor for the synthesis of nitric oxide (NO), reduces healing time of injuries (particularly bone), quickens repair time of damaged tissue and decreases blood pressure.


Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.


Soluble in water (50 mg/ml).


Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₄N₄O₂
Schmelzpunkt: 244 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002635
CAS-Nummer: 74-79-3

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=1.6, 6N HCl) +26.3° to +27.7°
Moisture ≤0.3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR