, Pulver

Lieferant: MP Biomedicals

Synonyme: pNPP, ortho-Phosphorsäure(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, 4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, p-Nitrophenylphosphat Dinatriumsalz Hexahydrat, ortho-Phosphorsäuremono(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, Phosphatasesubstrat, 4-Nitrophenyldihydrogenphosphat Dinatriumsalz Hexahydrat

0210087825 0210087850 02100878.5 0210087801 0210087805 0210087810
ICNA0210087825EA 249 EUR
ICNA0210087825 ICNA0210087850 ICNA02100878.5 ICNA0210087801 ICNA0210087805 ICNA0210087810
, Pulver
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)

p nitrophenol is the substrate of choice for use with alkaline phosphatase in ELISA procedures. This substrate produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 M NaOH and read at 405 nm.


p-nitrophenol is suitable for use as a substrate for alkaline and acid phosphatase.


Typically a pNPP concentration of 1 mg/ml is used


Soluble in water.

Formel: C₆H₄NNa₂O₆P·6H₂O
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Water Content 27 - 31%
Free p-Nitrophenol ≤0,005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR