6-Benzylaminopurin, Pulver

Lieferant: MP Biomedicals
Achtung

02100912.5 0210091205 0210091225
ICNA02100912.5EA 24.5 EUR
ICNA02100912.5 ICNA0210091205 ICNA0210091225
6-Benzylaminopurin, Pulver
6-Benzylaminopurin

6-Benzylaminopurine is a plant cell division factor (a plant growth factor).


  • E (270) = ~19,054 (methanol) (Lit.)


6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses. BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg′s medium, and Chu′s N6 medium. Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables.


BAP is an inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.


E (270) = ~19,054 (methanol) (Lit.)


Soluble in glacial acetic acid (50 mg/ml). Soluble in concentrated ammonium hydroxide, 0,1 N Sodium hydroxide (10 mg/ml - slightly hazy and yellowish solution), methanol, or dioxane.


Bitte beachten Research Use Only (RUO).

Formel: C₁₂H₁₁N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00005572
CAS-Nummer: 1214-39-7

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 226 - 236 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR