Bile sac, powder, MP Biomedicals
Lieferant: MP Biomedicals
0210100980
ICNA0210100980EA
101
EUR
ICNA0210100980
Bile sac, powder, MP Biomedicals
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
Bile Sac Powder is Gall Bladder powder.
- CAS: 8008-63-7
Foreign activity: Salmonella negative, Escherichia coli negative