Stearinsäure ≥99%, weißes kristallines Pulver
Lieferant: MP Biomedicals
Stearic acid, an anti-inflammatory agent, is the hydrogenation product of linoleic acid.
Stearic acid has been shown to induce a decrease of cell growth and adhesion and increase hydrophobic properties of collagen films. Stearic Acid has been noted to induce an increase in ICAM-1 expression in cells, via an increase in phosphorylation of the transcriptional regulator NFκB.
Soluble in alcohol, benzene, chloroform, acetone, carbon tetrachloride, carbondisulphide; very slightly soluble in water.
Formel:
C₁₈H₃₆O₂ |
MDL:
MFCD00002752 CAS-Nummer: 57-11-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...