2,6-Di-tert-butyl-p-kresol, powder_crystalline

Lieferant: MP Biomedicals

Synonyme: DBPC, Butylhydroxytoluol, 2,6-Di-tert-butyl-4-methylphenol, Ionol, BHT

0210116280 0210116290 0210116291
ICNA0210116280EA 23.9 EUR
ICNA0210116280 ICNA0210116290 ICNA0210116291
2,6-Di-tert-butyl-p-kresol, powder_crystalline
2,6-Di-tert-butyl-p-kresol

Butylated hydroxytoluene is a phenolic antioxidant.


Butylated hydroxytoluene (BHT) has been shown to inhibit lipid peroxidation. It causes lung injury and promotes tumors in mice, but this may be due to a metabolite of BHT, 6-tert-butyl-2-[2′-(2′-hydroxymethyl)-propyl]-4-methylphenol. Metabolites of BHT have also been reported to induce DNA strand breaks and internucleosomal DNA fragmentation (a characteristic of apoptosis) in cultured cells. In rats, a single intraperitoneal injection of BHT (60 mg/kg body mass) results in a significant increase in nuclear DNA methyl transferase activity in the liver, kidneys, heart, spleen, brain and lungs. Incubation of alveolar macrophages with BHT significantly reduced the level of TNF-α which may explain the mechanism by which this antioxidant reduces inflammation. Preincubation of aspirin-treated platelets with BHT inhibits the secretion, aggregation, and protein phosphorylation induced by protein kinase C activators. BHT was also found to inhibit the initiation of hepatocarcinogenesis by aflatoxin B1.


Insoluble in water; freely soluble in toluene, soluble in methanol, ethanol (100 mg/ml), isopropanol, methyl ethyl ketone, acetone, Cellosolve, benzene and most other hydrocarbon solvents. More soluble in food oils and fats than butylated hydroxyanisole.


Bitte beachten Research Use Only (RUO).

Formel: C₁₅H₂₄O
Molecular Weight: 220.35 g/mol
Siedepunkt: 265
Flammpunkt: 127
Lagertemperatur: Raumtemperatur
MDL: MFCD00011644
CAS-Nummer: 128-37-0
Merck Index: 13,01547

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 68 - 74 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR