ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4) ≥98%

Lieferant: MP Biomedicals

Synonyme: Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz (reduzierte Form)

0210116725 0210116750 0210116780 0210116783 0210116791
ICNA0210116725EA 126 EUR
ICNA0210116725 ICNA0210116750 ICNA0210116780 ICNA0210116783 ICNA0210116791
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4) ≥98%
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4)

Prepared by chemical reduction of NADP.


  • White powder


Store at −0 °C, desiccate, protect from light, store under nitrogen

Formel: C₂₁H₂₆N₇Na₄O₁₇P₃
Molecular Weight: 833.35 g/mol
MDL: MFCD00036263
CAS-Nummer: 2646-71-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR