Buttersäure ≥99.5%

Lieferant: MP Biomedicals
Gefahr

0210116980
ICNA0210116980EA 23.9 EUR
ICNA0210116980
Buttersäure ≥99.5%
Buttersäure

Butyric acid is used in the preparation of various butyrate esters. Low-molecular-weight esters of butyric acid, such as methyl butyrate, have mostly pleasant aromas or tastes. As a consequence, they find use as food and perfume additives. It is also used as an animal feed supplement, due to the ability to reduce pathogenic bacterial colonization.

Formel: C₄H₈O₂
MDL: MFCD00002814
CAS-Nummer: 107-92-6
UN: 2820
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR