1-Hexadecanol

Lieferant: MP Biomedicals

0210130990 0210130980
ICNA0210130990EA 92.1 EUR
ICNA0210130990 ICNA0210130980
1-Hexadecanol
1-Hexadecanol

Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.


Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.


Easily soluble in diethyl ether, soluble in acetone, chloroform; slightly soluble in alcohol.


Store at +4 °C

Formel: H₃C(CH₂)₁₄CH₂OH
Siedepunkt: 305…330 °C (1013 hPa)
Schmelzpunkt: 46…52 °C
Dichte: 0,81 g/cm³ (20 °C)
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004760
CAS-Nummer: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR