CAPS (3-(Cyclohexylamino)propansulfonsäure), weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 3-Cyclohexylaminopropanesulphonic acid

0210143525 0210143580 0210143583 0210143591
ICNA0210143525EA 35 EUR
ICNA0210143525 ICNA0210143580 ICNA0210143583 ICNA0210143591
CAPS (3-(Cyclohexylamino)propansulfonsäure), weißes Pulver

CAPS is a zwitterionic biological buffer which has minimal reactivity with enzymes or proteins and minimal salt effects. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8,5.


  • ΔpK/ΔT: - 0,0322


CAPS has been used to study pH-dependent processes in enzymology and as an eluent buffer component in the separation of basic drugs by HPLC.


Very soluble in water (1 M - clear, colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₁NH(CH₂)₃SO₃H
Molecular Weight: 221.32 g/mol
Schmelzpunkt: 324
MDL: MFCD00003837
CAS-Nummer: 1135-40-6
Merck Index: 13,01773

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 5,0 - 6,3
pka 10,4 (25 °C) (Lit.)
Refractive Index 1,52
Sterilisation of Solutions Autoclave


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR