L(+)-Cystein Hydrochlorid Monohydrat, Kristalle

Lieferant: MP Biomedicals

0210144605 02101446.1 02101446.3 02101446.5 0210144601
ICNA0210144605EA 2037 EUR
ICNA0210144605 ICNA02101446.1 ICNA02101446.3 ICNA02101446.5 ICNA0210144601
L(+)-Cystein Hydrochlorid Monohydrat, Kristalle

Cysteine is a major biological source of sulphur and is one of the two common sulphur-containing amino acids. It is a NMDA glutamatergic receptor antagonist.


Freely soluble in water (50 mg/ml).

Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065606
CAS-Nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR